Nunc™ EasYDish™ Cell Culture Dishes with Nunclon™ Supra Surface
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
150470 150472 150474 150476 150478
734-3641EA 7.2 EUR
734-3641 734-3638 734-3643 734-3636 734-3639
Nunc™ EasYDish™ Cell Culture Dishes with Nunclon™ Supra Surface
Cultuurschalen Celcultuurschalen
Experience optimal performance with Thermo Scientific Nunc™ EasYDish™ dishes with Nunclon™ Supra surface treatment for culturing a wide variety of fastidious adherent cells.
  • The beveled grip ring is easy to hold and stays secure in your gloved hand
  • Available in four sizes: 35, 60, 100, and 150 mm
  • Sterile, not autoclavable
  • Clear, round, flat bottom, not gridded
  • Single use
  • Consistent lot-to-lot performance
  • Non-cytotoxic and non-pyrogenic
  • Biocompatible per USP Class VI for raw materials
  • 5 years of shelf life
Intended for general cell culture applications,More Product Information