ISO Safety Signs - Warning; Substance or Mixture Presenting a Health Hazard
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
309769 309770
BRDY309769EA 6.06 EUR
BRDY309769 BRDY309770
ISO Safety Signs - Warning; Substance or Mixture Presenting a Health Hazard
Pictogrammen
ISO safety sign depicts 'Warning; Substance or mixture presenting a health hazard' message. Avoiding any possibility to have contact with or spills of substances or mixtures considered hazardous to health.
  • Triangle shape
  • Black on yellow finish
  • 0,099 mm thickness
  • Permanent acrylic adhesive and glossy finish
Applications includes facility identification, hazard communication identification.
Certificaten: Compliant with Directive 2011/65/EU and 2015/863.