Powder funnel with NS core
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1710-080-29100 1710-02-080241 1710-100-29120
NEUB1710-080-29100EA 28.8 EUR
NEUB1710-080-29100 NEUB1710-02-080241 NEUB1710-100-29120
Powder funnel with NS core
Trechters Poedertrechters
Borosilicate glass 3,3.
  • Chemically resistant glass
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Quick and easy to clean
Ideal for transferring powders, can also be used for liquids.