ORP electrode, TA-SensoLyt PtA
Leverancier: WTW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WTWA103711EA 424 EUR
WTWA103711
ORP electrode, TA-SensoLyt PtA
Elektroden Redox Electrodes
ORP combination electrode (platinum) for installation in depth armature series TA 197-pH, polymer electrolyte, pin hole junction.