Stainless Steel Shoe and Garment Storage Cupboards
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CRBS-SC10P CRBS-SC20P
TEKMCRBS-SC10PEA 0 EUR
TEKMCRBS-SC10P TEKMCRBS-SC20P
Stainless Steel Shoe and Garment Storage Cupboards
Meubilair Omkleedrekken
These stainless steel shoe storage cupboards are manufactured from robust, easy to clean, 304L-grade stainless steel and are strong and sturdy due to the stainless steel adjustable feet. The sloping top increases the hygienic quality of the shoe storage compartment and pre-drilled drainage holes encourages easy cleaning.
  • 304L-grade stainless steel
  • Brushed dull polish finish
  • Freestanding
  • Sloping top
This shoe storage cupboard ensure that employees can store their footwear safely and in an organised manner, ensuring quick dress-up/dress-down within a changing room.