Hygienox Lecterns with Tubular Rear Tie Bar
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TLLEHYEPR6040 TLLEHYEPRR6040
TEKMTLLEHYEPR6040EA 0 EUR
TEKMTLLEHYEPR6040 TEKMTLLEHYEPRR6040
Hygienox Lecterns with Tubular Rear Tie Bar
Meubilair Tafels
The Hygienox lectern with tubular rear tie bar is designed specifically for pharmaceutical and cleanroom environments.
  • 316L-grade stainless steel
  • Electropolished (with an ultra-hygienic <0,3 RA finish)
  • Open corners on lectern tops and tubular legs
  • Inclined top with lip
  • 4× Hygienox 316 stainless steel feet with no exposed thread (Static)
  • 4× Hygienox 316 polyethylene castors (2 braked) (Mobile)
  • 100 kg payload
Hygienox 316L-grade stainless steel lecterns are made specifically for cleanroom environments, giving extreme protection against pitting and crevice corrosion caused by harsh clean down routines (such as chlorine and hydrogen peroxide).
Manufactured in electropolished 316L-grade stainless steel for an ultra-hygienic environment, it comes with a smoother <0,3 RA finish. Easy clean features include no horizontal ledges, from the tubular rear tie bar to fully sloping writing area.
The intelligent design and specifications of the hygienox lecterns helps to not only reduce cleandown times, but also improves productivity. The 600 mm width is ideal for holding documents up to A3 in size, while the front ledge keeps documents in place.