PE Medium Adhesion Cleanroom Tape
Leverancier: ULTRATAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1154WH038-P3D 112GR075-P3T 112RE075-P3T 112YE075-P3T
UTAP1154WH038-P3DEA 325 EUR
UTAP1154WH038-P3D UTAP112GR075-P3T UTAP112RE075-P3T UTAP112YE075-P3T
PE Medium Adhesion Cleanroom Tape
Tape
Particle-free, residue-free tapes.
  • Medium adhesion polyethylene tapes
  • Good alternative to vinyl tapes
  • UltraClean material
These general use and cleanroom construction tapes bond quickly to all surfaces. The adhesive strength is virtually identical to vinyl tapes. The polyethylene backing can be written on with a cleanroom pen, and it can be torn by hand.
UltraTape 1154/0154 is also recommended for in-process applications due to its low outgassing and metal properties. UltraTape 1112 has the same adhesive system (but different ratio of ingredients), and a medium density as opposed to a low density polyethylene backing.