Axygen® Transport Tubes with Screw Caps, Self Standing
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
331-77-202 331-77-252
AXYG331-77-202EA 114 EUR
AXYG331-77-202 AXYG331-77-252
Axygen® Transport Tubes with Screw Caps, Self Standing
Buisjes Reageerbuisjes
Secure fit between tube and cap to give leak proof seal.
  • Non pyrogenic
  • Large frosted writing area