Chlorine (Free and Total) Reagent Set
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
540210 530210
LOVI540210EA 72.2 EUR
LOVI540210 LOVI530210
Chlorine (Free and Total) Reagent Set
Milieutestkits
Chlorine (Free and Total) Reagent set are used in water analysis.
  • Cut Cost, not quality - proven results at competitive prices
  • Supplies enough Reagent for 30 days of continuous operation
  • Designed to fit directly into the Hach® CL17™
  • Comparable chemistries and bottle sizes for ease of use
Leveringsinformatie: Reagents set in bag: 1 bottle DPD indicator solution 'free chlorine' (473 ml), 1 bottle DPD buffer solution 'free chlorine' (473 ml), 1 bottle DPD indicator powder.