Chucks for RZR Series Electronic Stirrers
Leverancier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
509-02000-00 509-01000-00-0
441-1202EA 762 EUR
441-1202 HEID509-01000-00-0
Chucks for RZR Series Electronic Stirrers
Roerders Roergereedschap
Chucks are designed for use with all RZR series overhead electronic stirrers.
The chuck and key set features a hard chrome finish. The flexible shaft features a maximum torque of 150 Ncm.