VWR® Jar for Anaerobic Cultures
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
499240OOEA 208 EUR
499240OO 499244OO
VWR® Jar for Anaerobic Cultures
Anaerobe systemen
This jar is designed for anaerobic, microaerophilic and or different concentrations of CO₂ cultures. It can be used with any sachet to generate special atmospheres and to contain up to 12 plates.
  • No catalyst required
  • Metal cover subjected by a threaded closure pressure
  • NBR O-ring
Two colours of lid available (red or blue) to easily identify different incubation atmospheres inside the jars during use.