DURAN® Desiccators, Mobilex without Porcelain Plate
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
247866103 247866909
SCOT247866103EA 239 EUR
SCOT247866103 SCOT247866909
DURAN® Desiccators, Mobilex without Porcelain Plate
Exsiccatoren Exsiccatoren, glas
DURAN® clear borosilicate glass 3.3, vacuum desiccator with Mobilex screw thread without porcelain plate.