Duran® Glass Cones with Ridge, With Tip
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286192905 286191909
SCOT286192905EA 124 EUR
SCOT286192905 SCOT286191909
Duran® Glass Cones with Ridge, With Tip
Geslepen Geslepen hulzen
Rigid glass connections and the most frequently used connection type in glass apparatus.
  • Exact cone angles for excellent quality
  • Low joint surface roughness
  • With or without tip
  • Provides tight seals
Certificaten: Acc. to DIN 12 249.