Ice Maker, HC Series
Leverancier: HOSHIZAKI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FM-120KE-HC FM-120KE-HCN
HOSHFM-120KE-HCEA 5845 EUR
HOSHFM-120KE-HC HOSHFM-120KE-HCN
Ice Maker, HC Series
IJsmakers
The FM-120KE-HC/HCN are self-contained ice makers, which utilise natural refrigerant R290.
  • Automatic flush system to reduce sediment build up
  • Stainless steel exterior with integrated door handles provides a hygienic and high quality finish
  • Member of the Emerald class,Hoshizaki’s range of climate friendly ice makers
  • Quality of ice is always consistent
Closed water circuit offers the ultimate contamination protection,by reducing the number of points at which impurities can enter the ice making process.
Electronically controlled to ensure an optimised ice making process under varying circumstances without the need for physical adjustments.
Easily cleanable air filter allows end-users to carry out routine cleaning schedules. Extends product life expectancy and reduces the frequency and costs of maintenance call-outs.