Reprospher 100 Diol HPLC Column
Leverancier: DR MAISCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RS13.D0.S1003 RS15.D0.S2520
MAISRS13.D0.S1003EA 334 EUR
MAISRS13.D0.S1003 MAISRS15.D0.S2520
Reprospher 100 Diol HPLC Column
Kolommen voor chromatografie
  • Surface area: 350 m²/g
  • PH range: 2 to 8