L-Phenylalanine
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
32191.01 32191.02 32191.03
SERA32191.01EA 0 EUR
SERA32191.01 SERA32191.02 SERA32191.03
L-Phenylalanine
L-Phenylalanine
Formule: C₉H₁₁NO₂
Kookpunt: 313 °C (1013 hPa)
Smeltpunt: 173 °C
Dichtheid: 0,754 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064227
CAS nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271