L-Phenylalanine, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.07267.1000 1.07267.9010
1.07267.1000EA 1060 EUR
1.07267.1000 1.07267.9010
L-Phenylalanine, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
L-Phenylalanine
Formule: C₉H₁₁NO₂
Kookpunt: 313 °C (1013 hPa)
Smeltpunt: 173 °C
Dichtheid: 0,754 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064227
CAS nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271