L-Phenylalanine, Millipore®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
M-Clarity™ quality level = MQ100
1.07256.0100 1.07256.1000 1.07256.9010
1.07256.0100EA 116 EUR
1.07256.0100 1.07256.1000 1.07256.9010
L-Phenylalanine, Millipore®
L-Phenylalanine
Formule: C₉H₁₁NO₂
Kookpunt: 313 °C (1013 hPa)
Smeltpunt: 173 °C
Dichtheid: 0,754 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064227
CAS nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271
Order Now
Specificaties
Environmentally Preferable