L-Phenylalanine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4031148.0250 4031148.1000
BACE4031148.0250EA 0 EUR
BACE4031148.0250 BACE4031148.1000
L-Phenylalanine
L-Phenylalanine
Formule: C₉H₁₁NO₂
Kookpunt: 313 °C (1013 hPa)
Smeltpunt: 173 °C
Dichtheid: 0,754 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064227
CAS nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR