2,4,6-Trimethylbenzonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00289.1 TL00289.10 TL00289.50
MAYBTL00289.1EA 0 EUR
MAYBTL00289.1 MAYBTL00289.10 MAYBTL00289.50
2,4,6-Trimethylbenzonitrile
2,4,6-Trimethylbenzonitrile
Formule: C₁₀H₁₁N
MDL Nummer: MFCD00016378
CAS nummer: 2571-52-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR