4-Nitro-m-toluonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00730.1 TL00730.10 TL00730.50
MAYBTL00730.1EA 0 EUR
MAYBTL00730.1 MAYBTL00730.10 MAYBTL00730.50
4-Nitro-m-toluonitrile
4-Nitro-m-toluonitrile
Formule: C₈H₆N₂O₂
Smeltpunt: 80 °C
MDL Nummer: MFCD00017615
CAS nummer: 96784-54-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR