4'-Nitroacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01771.1 SB01771.10 SB01771.50
MAYBSB01771.1EA 0 EUR
MAYBSB01771.1 MAYBSB01771.10 MAYBSB01771.50
4'-Nitroacetophenone
4'-Nitroacetophenone
Formule: C₈H₇NO₃
Kookpunt: 275 °C (1013 hPa)
Smeltpunt: 76…78 °C
Vlampunt: 149 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007355
CAS nummer: 100-19-6
EINECS: 202-827-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR