DL-α-Phenylalanine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-DL-Phe-OH, DL-Phenylalanine, 3-Phenyl-DL-alanine

4031150.0250 4031150.1000
BACE4031150.0250EA 0 EUR
BACE4031150.0250 BACE4031150.1000
DL-α-Phenylalanine
DL-α-Phenylalanine
Formule: C₉H₁₁NO₂
MW: 165.19 g/mol
Kookpunt: 313
Smeltpunt: 173
MDL Nummer: MFCD00064225
CAS nummer: 150-30-1
EINECS: 205-756-7
Merck Index: 13,07355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR