2-Bromoisobutyric acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Bromo-2-methylpropanoic acid, α-Bromoisobutyric acid, 2-Bromo-2-methylpropionic acid
305201000 305200050 305205000
ACRO305201000EA 61.3 EUR
ACRO305201000 ACRO305200050 ACRO305205000
2-Bromoisobutyric acid 98%
2-Bromoisobutyric acid
Formule: C₄H₇BrO₂
Kookpunt: 198…200 °C (760 mmHg)
Smeltpunt: 41…46 °C
Dichtheid: 1,43 g/cm³ (20 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004168
CAS nummer: 2052-01-9
EINECS: 218-139-2
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow to orange Semitransparent crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
Melting point 41 to 46 °C
Titration with NaOH ≥97.5 %