Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CKM12-12F-12F CKM12-12K-12K
ENTECKM12-12F-12FEA 0 EUR
ENTECKM12-12F-12F ENTECKM12-12K-12K
Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Ventielen Check Valves
Corrosion-resistant Galtek® check valves reduce contamination potential in your high-purity chemical applications and provide clean delivery of sophisticated chemicals.
  • O-ring free design reduces potential contamination
  • Simple, clean, compact valve provides high flow chemical delivery
  • No metal parts - will not corrode or contribute to contamination
  • Poppet design allows a high flow through rate and maximizes rinsability
An O-ring-free design provides a simple, clean, compact valve with a high flow rate and increased reliability. All wetted parts are molded PFA and machined PTFE and provides corrosion resistence and a cleaner overall process.
Applications include bulk chemical distribution, wet processing and chemical dispensing.