Borane dimethyl sulphide complex
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Packaged under argon in resealable ChemSeal™ bottles.
Packaged under Argon in resealable ChemSeal® bottles
CAS No.: 13292-87-0
Molecular Formula: C2H9BS
Formula Weight: 75.97
Flash Point: -1°(31°F)
Physical Form: Liquid
Density: 0.801
Refractive Index: 1.4570
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless to pale yellow
Odour: Unpleasant
Melting Point: -38°
Boiling Point: ca 44° dec.
Beilstein Reference: 3663489
UN No.: 2924
MDL No.: MFCD00013189
Formule:
C₂H₉BS Kookpunt: ∼44 °C (ontbinding) Smeltpunt: –38 °C Dichtheid: 0,801 Vlampunt: –1 °C (31 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013189 CAS nummer: 13292-87-0 EINECS: 236-313-6 UN: 2924 ADR: 3,II |