Cesium hydroxide 50% (w/w) , 99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
16505.MZEA 366 EUR
16505.MZ 16505.NF
Cesium hydroxide 50% (w/w) , 99% (metals basis)
Cesium hydroxide
Purity: 99% CAS No.: 21351-79-1 Molecular Formula: CsOH Formula Weight: 149.91 Physical Form: Liquid Density: 1.725 Appearance: Colourless Odour: Odourless UN No.: 2681 MDL No.: MFCD00010964
Formule: CsOH
MW: 149.91 g/mol
Smeltpunt: 338.85
Dichtheid: 1.725
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010964
CAS nummer: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8 II