Chloramphenicol

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

16785.01 16785.02
SERA16785.01EA 0 EUR
SERA16785.01 SERA16785.02
Chloramphenicol
Chloramphenicol
Formule: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Kookpunt: 645 °C (1013 hPa)
Smeltpunt: 150 °C
Dichtheid: 1,49 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00078159
CAS nummer: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR