SymLine Automatic Switching for Container
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
108334-T 107167 107165 106741 106732 107168 107166 399204 108335-T 108336-T 117111 307338 307380 307448 307970 307971 318958 399203 106751 106752 106733 106735
SCTV108334-TEA 215 EUR
SCTV108334-T SCTV107167 SCTV107165 SCTV106741 SCTV106732 SCTV107168 SCTV107166 SCTV399204 SCTV108335-T SCTV108336-T SCTV117111 SCTV307338 SCTV307380 SCTV307448 SCTV307970 SCTV307971 SCTV318958 SCTV399203 SCTV106751 SCTV106752 SCTV106733 SCTV106735
SymLine Automatic Switching for Container
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The ready-to-use automatic switching sets prevent the overflowing of disposal containers and the associated leakage of hazardous, toxic, or explosive liquid waste.
  • Coordinated system for monitoring the safe filling of disposal containers
  • For connection to two disposal containers (not included in the set)
  • Set consists of Built-in Signalbox2, SwitchBox, ball valve, and two signal cables
  • Optionally available as an ATEX version with an additional switch amplifier for two channels
Expandable sets for safe fill level monitoring during disposal in containers.