Blotting Paper for Cobb Test, Grade A 250
Leverancier: Ahlstrom-Munksjö
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.412.210210 3.412.200 3.412.180
BINZ2.412.210210EA 86.9 EUR
BINZ2.412.210210 BINZ3.412.200 BINZ3.412.180
Blotting Paper for Cobb Test, Grade A 250
Vloeipapier
Special absorbent paper suitable for the determination of absorption of water in the production of sized paper according to the Cobb method and testing the formation of sheets in the cellulose paste industry.
  • Basis weight 250 g/m²
  • Water absorption capacity 450 g/m²
  • Capillary rise, MD 70 mm/10 min
Certificaten: For Cobb test according to DIN EN 20535 or ISO 535. This product is manufactured in conformance with the quality management standard DIN EN ISO 9001.