WHEATON® Individually Wrapped Sterile Dippers
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
327131 327130 327136 327135 360240 360239
WHEA327131EA 179 EUR
WHEA327131 WHEA327130 WHEA327136 WHEA327135 WHEA360240 WHEA360239
WHEATON® Individually Wrapped Sterile Dippers
Monsternemers Monsterdompelaars
Ideal for safe sample collection and transportation in a single container, eliminates the risk of cross-contamination.
  • Handle neatly snaps off after sample collection for ease of transportation
  • Long handle length accommodates a majority of sampling applications
  • Choice of clear and blue polystyrene
  • Sterilised by gamma irradiation
  • Supports food industry and pharmaceutical requirements
Certificaten: Tested in accordance with BS EN 14254 for safe, leak-free performance.