Basket Trolleys
Leverancier: Hupfer Metallwerke
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HUPF0128892EA 1210 EUR
HUPF0128892 HUPF0128912
Basket Trolleys
Transportwagens
Basket transport trolleys in high version for empty and loaded 500×500 mm crockery baskets.
  • Made entirely of high-quality stainless steel
  • Robust, self-supporting and hygienic design
Tubular frame rack consisting of square tubes with plastic end caps and welded L-shaped shelf rails for holding 500×500 to 515×515 mm. Push-through protections on both sides as per DIN EN 18867-2. 4 polyethylene disc bumpers serve as bumpers and protect trolley on all sides as well as building-side walls from being damaged.
Trolleys runs on four swivel casters of which two with total locks, Ø 125 mm with pin fastening. The Hupfer basket transport trolley has outstanding handling properties due to its very low empty weight, as well as a very large usable height between the shelf rails of 145 mm.