VWR® Hitachi UH4150 UV-VIS/NIR Spectrophotometers
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1J1-0013 1J1-0003
634-0951EA 53500 EUR
634-0951 634-0952
VWR® Hitachi UH4150 UV-VIS/NIR Spectrophotometers
Spectrofotometers en fotometers UV/Vis spectrofotometers
These UV-Vis/NIR spectrophotometers offer enormous flexibility for the evaluation and quality control of solid samples, typically optical components or coated surfaces. They can be configured for non destructive measurements when equipped with relevant accessories for applications such as:
  • Micro sample reflectance
  • Wafer reflectance/transmission
  • Optical thin film reflection
  • Lens transmittance
  • Solar cells and coated glass evaluation
High precision accessories can measure at a range of incident angles.
For coated glass surfaces, the variable angle absolute reflectance accessory (20 to 60°) measures the absolute reflectance and transmittance at a desired angle by rotating the detector and the sample stage independently.
Bestelinformatie: For more information on the range of accessories, please contact your local Avantor agency.