WHEATON® Wide-Mouth Packers, Amber
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W217960 W217961
WHEAW217960EA 267 EUR
WHEAW217960 WHEAW217961
WHEATON® Wide-Mouth Packers, Amber
Flessen Glazen flessen
Wheaton wide-mouth packers are manufactured from amber soda lime glass that conforms to USP Type III requirements.
Amber glass provides protection for light sensitive products. The wide-mouth design is ideal for use with dry powders. A partitioned carton protects bottles from scratches during shipping. Each case also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Bestelinformatie: Available with caps attached or in bulk packs without caps. Replacement screw caps can be purchased separately.