WHEATON® MicroLiter® μLplate® Snap Caps and μLmats™
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07-0045MN 07-0045N 07-0045B 07-0045G 07-0045R 07-0070G 07-0070R 07-0075B 07-0075N 07-0075R 07-0020R 07-0020N 07-0020B 07-0070MN 07-0075MN 07-0021N
WHEA07-0045MNEA 275 EUR
WHEA07-0045MN WHEA07-0045N WHEA07-0045B WHEA07-0045G WHEA07-0045R WHEA07-0070G WHEA07-0070R WHEA07-0075B WHEA07-0075N WHEA07-0075R WHEA07-0020R WHEA07-0020N WHEA07-0020B WHEA07-0070MN WHEA07-0075MN WHEA07-0021N
WHEATON® MicroLiter® μLplate® Snap Caps and μLmats™
Afdichtingen Afdichtingen van microtiterplaten
A comprehensive line of 96-well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials.
Polypropylene μLplates™ have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples can be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options let you choose the specifications appropriate for your application.
The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer you secure vessels to maintain even the most volatile samples.