Xonachips® Cell Culture Plates
Leverancier: LIEFERSERVICE UND LOGISTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
XC450 XC900
LIEFXC450EA 0 EUR
LIEFXC450 LIEFXC900
Xonachips® Cell Culture Plates
Microplaten Celcultuurplaten
XonaChips® are pre-assembled and pre-bonded, making them easier-to-use than our original silicone devices. XonaChips® are optically transparent and ideal for fluorescence live imaging. XonaChips® improve long-term culture of primary neurons and are especially useful for culturing human stem cell-derived neurons, providing better cell attachment and long-term growth.
  • Xona’s easiest-to-use compartmentalized platform made in cyclic olefin copolymer
  • Optically transparent and fully assembled
  • Improved long-term culture of human derived neurons
Two compartment XonaChips® have 150, 450, or 900 µm microgroove barriers (XC450, XC900). Each box of five XC Series XonaChips® includes a 2 ml tube of XC Pre-Coat™.