Proteinase K Solution
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROK-50 PROK-200
PROK-50EA 638 EUR
PROK-50 OMEGPROK-200
Proteinase K Solution
Enzymen
  • Concentration: 20 mg/ml
Opgelet: For research use only. Not for use in diagnostic procedures.
Order Now
Specificaties
Environmentally Preferable
Bron
Species
Learn more