WHEATON® Container, Wide Mouth
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W209916 W209912
WHEAW209916EA 180 EUR
WHEAW209916 WHEAW209912
WHEATON® Container, Wide Mouth
Monsternamepotjes Containers voor algemeen gebruik
Container made of polystyrene with foam PE lined polypropylene screw cap.
  • Clear polystyrene offers excellent calrity of contents within
  • Ideal for the storage of solid samples and powders
  • Good resistance to inorganic chemicals