SureSwab Rapid Multiple Drug Test Kit
Leverancier: SURESCREENTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
SureSwab rapid kit is quick and easy form of oral drug screening.
- Optimising collection time
- Smaller sponge for easier saturation
- Indicator to display sponge is fully saturated
- Detection of both parent and metabolic cannabis
- Slimline design for easier transportation
- Test up to six parameters simultaneously
The SureSwab Rapid is an easy-to-use screening test to detect multiple drug parameters in oral fluid. It is designed to be less invasive for improved donor experience and enables rapid collection time. A simple solution suitable for workplace, rehab, and clinical testing.
Leveringsinformatie: Supplied with test strip, swab collector, procedure card and product IFU.
Opgelet: For professional use only.