ExoBrite™ True EV Membrane Stains
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30129-T 30129
30129-TEA 196 EUR
30129-T 30129.
ExoBrite™ True EV Membrane Stains
Koppelings- en labelingssystemen voor eiwitten
Lipophilic membrane dyes designed specifically for EV research and the best choice for general pan-EV staining.
  • Superior alternative to common membrane dyes like PKH, DiO, DiI, DiD
  • Near-complete coverage of EVs in a sample
  • Broad compatibility with EVs isolated from different sources
  • Compatible with antibody-co-staining
ExoBrite™ True EV membrane stains demonstrate higher (near-complete) coverage of EVs over competitor membrane dyes in fluorescence nanoparticle tracking analysis (fNTA). The dyes also offer clear differentiation of EVs from non-specific particles during flow detection. ExoBrite™ True EV Membrane Stains labeled all EV types tested to date.