ESD Occupational Clogs, Tiefbett Leder BAD TÖLZ, 19380
Leverancier: SCHURR SCHUHVERTRIEB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
51938.01.14.43 51938.01.14.41 51938.01.14.38 51938.01.14.36 51938.01.14.37 51938.01.14.39 51938.01.14.40 51938.01.14.48 51938.01.14.47 51938.01.14.46 51938.01.14.45 51938.01.14.44 51938.01.14.42
SCRR51938.01.14.43EA 65.6 EUR
SCRR51938.01.14.43 SCRR51938.01.14.41 SCRR51938.01.14.38 SCRR51938.01.14.36 SCRR51938.01.14.37 SCRR51938.01.14.39 SCRR51938.01.14.40 SCRR51938.01.14.48 SCRR51938.01.14.47 SCRR51938.01.14.46 SCRR51938.01.14.45 SCRR51938.01.14.44 SCRR51938.01.14.42
ESD Occupational Clogs, Tiefbett Leder BAD TÖLZ, 19380
Schoenen en laarzen
Sandal with instep and heel straps, perforated, cork footbed.
  • Upper material: Leather
  • Sole material: Rubber
  • Weight: 295 g
Certificaten: EN ISO 20347:2004.