24(S),25-Epoxycholesterol ≥98% (by TLC)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Packaged under inert gas.
BMLGR2310001 BMLGR2310005
ENZOBMLGR2310001EA 245 EUR
ENZOBMLGR2310001 ENZOBMLGR2310005
24(S),25-Epoxycholesterol ≥98% (by TLC)
24(S),25-Epoxycholesterol
LXR ligand.
Endogenous agonist for the nuclear receptor LXR.
Soluble in ethanol (10 mg/ml) or DMSO (10 mg/ml).
Opgelet: Should be used immediately following dissolution. Store with desiccant.
Formule: C₂₇H₄₄O₂
Smeltpunt: 156…159 °C
CAS nummer: 77058-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c11fa0dcf085da13cdb6fb657808bb83018888f&vid=09cb0ef6a0e47d0914adacd42cb1207c1135fefc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity ≥98% (TLC)
Identity Determined by NMR