MK-571 sodium salt 90 HPLC_ASSAY_METHOD
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Sodium (E)-3-(((3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)((3-(dimethylamino)-3-oxopropyl)thio)methyl)thio)propanoate, MK-571 sodium
BMLRA1090005 BMLRA1090025
ENZOBMLRA1090005EA 86.9 EUR
ENZOBMLRA1090005 ENZOBMLRA1090025
MK-571 sodium salt 90 HPLC_ASSAY_METHOD
MK-571 sodium salt
A potent, selective, orally active leukotriene D₄ antagonist. Ki=2.1 nM for inhibition of [³H]-LTD₄ binding to human lung membranes. Competitively antagonizes contractions of guinea pig trachea and ileum and human trachea induced by LTD₄. Blocks LTD₄ and ascaris-induced bronchoconstriction in conscious squirrel monkeys.
Soluble in water (20 mg/ml), aqueous buffers, 100% ethanol (10 mg/ml) or DMSO.
Formule: C₂₆H₂₆ClN₂O₃S₂·Na
MW: 537 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11114392
CAS nummer: 115103-85-0
Order Now
Specification Test Results
Purity ≥90% (HPLC)