PP242 hydrate ≥99% (by HPLC)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZCHM1320001 ENZCHM1320005
ENZOENZCHM1320001EA 89.1 EUR
ENZOENZCHM1320001 ENZOENZCHM1320005
PP242 hydrate ≥99% (by HPLC)
PP242 hydrate
Inhibitor of both mTORC1 and mTORC2. Induces cell death in human and murine leukemia cells.
Inhibitor of the active site of mTOR kinase in both mTORC1 and mTORC2. pp242 has been shown to cause the death of certain human and murine leukemia cells more potently than rapamycin.
Formule: C₁₆H₁₆N₆O·H₂O
CAS nummer: 1092351-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=874173a617edd1561546100233df8e71ca79934e&vid=9a8989925161488f51730d28bb7e1dbbf5ea0846&prid=web&clid=SAPDAM