Niltubacin
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLGR3630100 BMLGR3630500
ENZOBMLGR3630100EA 89.1 EUR
ENZOBMLGR3630100 ENZOBMLGR3630500
Niltubacin
Niltubacin
HDAC inhibitor
Synthetic.
Negative control for Tubacin (tubulin acetylation inducer), a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6. Concentration of niltubacin in cell culture experiments typically ranges from 2-20µM.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db0b9eab71d11e3da3861925bb681c3aef1f3061&vid=f4fbc261087379ca1782a78aa7b62a619c63a350&prid=web&clid=SAPDAM