L-NIL dihydrochloride ≥97%
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX270010M010 ALX270010M050
ENZOALX270010M010EA 70.4 EUR
ENZOALX270010M010 ENZOALX270010M050
L-NIL dihydrochloride ≥97%
L-NIL dihydrochloride
NOS inhibitor
Selective inhibitor of inducible nitric oxide synthase (iNOS; NOS II). Shows anti-inflammatory activity.
CAS nummer: 159190-45-1