Fluvastatin sodium salt ≥98% (by HPLC)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: XU 62-320, rel-(3S,5R,E)-7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic acid sodium salt, Sodium rel-(3R,5S,E)-7-(3-(4-fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoate
ALX270466M010 ALX270466M050
ENZOALX270466M010EA 79.2 EUR
ENZOALX270466M010 ENZOALX270466M050
Fluvastatin sodium salt ≥98% (by HPLC)
Fluvastatin sodium salt
Anticholesterol agent. HMG-CoA inhibitor.
Anticholesterol agent. HMG-CoA reductase inhibitor and antioxidant.
Synthetic.
Formule: C₂₄H₂₅FNNaO₄
MW: 433.46 g/mol
MDL Nummer: MFCD00929076
CAS nummer: 93957-55-2