Sorafenib (BAY 43-9006) ≥95% (by HPLC)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, Sorafenib, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)phenoxy)-N-methylpicolinamide
ALX270469M001 ALX270469M005 ALX270469M025
ENZOALX270469M001EA 101 EUR
ENZOALX270469M001 ENZOALX270469M005 ENZOALX270469M025
Sorafenib (BAY 43-9006) ≥95% (by HPLC)
Sorafenib (BAY 43-9006)
Broad-spectrum kinase inhibitor
Broad-spectrum kinase inhibitor.
Synthetic.
Formule: C₂₁H₁₆ClF₃N₄O₃
MW: 464.82 g/mol
MDL Nummer: MFCD06411450
CAS nummer: 284461-73-0