Sinefungin 95 HPLC_ASSAY_METHOD
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX380070M001 ALX380070M005
ENZOALX380070M001EA 154 EUR
ENZOALX380070M001 ENZOALX380070M005
Sinefungin 95 HPLC_ASSAY_METHOD
Sinefungin
Methyltransferase inhibitor
Strong competitive inhibitor of methyltransferases which uses S-adenosyl-1-methionine as the methyl group donor to yield methylated products such as 5-methylcytosine or N6-methyl adenosine on DNA and RNA. In addition, sinefungin is involved in a number of physiological processes.
CAS nummer: 58944-73-3