(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: GSK 3 Inhibitor IX, BIO, 6BIO, 6-Bromoindirubin-3'-oxime, (2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one
ALX430156M001 ALX430156M005
ENZOALX430156M001EA 99 EUR
ENZOALX430156M001 ENZOALX430156M005
(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%
(2'Z,3'E)-6-Bromoindirubin-3'-oxime
Inibitor of phosphoinositide-dependent kinase 1 and GSK
Inhibitor of phosphoinositide-dependent kinase 1 (PDK1). Potent, reversible and ATP-competitive inhibitor of glycogen synthase kinase-3alpha/beta (GSK-3alpha/beta) (IC50=5nM).
Formule: C₁₆H₁₀BrN₃O₂
MW: 356.18 g/mol
MDL Nummer: MFCD08705318
CAS nummer: 667463-62-9